Skip to Content
MilliporeSigma
Codeine-6-β-D-glucuronide solution 1 mg/mL in methanol: water (2:8), ampule of 1 mL, certified reference material, Cerilliant®

Codeine-6-β-D-glucuronide solution

Codeine-6-β-D-glucuronide solution 1 mg/mL in methanol: water (2:8), ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C24H29NO9
CAS Number:
Molecular Weight:
475.49

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Quality Segment
Application
Package Size
Reference Material Type
Special Grade
Technique