Skip to Content
MilliporeSigma
Cobalt(II) nitrate hexahydrate JIS special grade, ≥98.0%

Cobalt(II) nitrate hexahydrate

Cobalt(II) nitrate hexahydrate JIS special grade, ≥98.0%
Synonym(s):
Cobaltous nitrate, Cobaltous nitrate hexahydrate, Nitric acid, cobalt(II) salt, Cobalt(II) nitrate hexahydrate
Linear Formula:
Co(NO3)2 · 6H2O
CAS Number:
Molecular Weight:
291.03

Products

Product Category
Brand
Sustainability
Color
Feature
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Reagent Type
Core
Special Grade