Skip to Content
MilliporeSigma
Cobalt(II) acetylacetonate ≥99.0% (KT)

Cobalt(II) acetylacetonate

Cobalt(II) acetylacetonate ≥99.0% (KT)
Synonym(s):
2,4-Pentanedione cobalt(II) derivative, Bis(2,4-pentanedionato)cobalt, Co(acac)2, Cobaltous acetylacetonate
Linear Formula:
Co(C5H7O2)2
CAS Number:
Molecular Weight:
257.15
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing