Skip to Content
MilliporeSigma
Cinchonine monohydrochloride hydrate 99%

Cinchonine monohydrochloride hydrate

Cinchonine monohydrochloride hydrate 99%
Synonym(s):
Cinchonan-9(S)-ol monohydrochloride
Empirical Formula (Hill Notation):
C19H22N2O · HCl · xH2O
CAS Number:
Molecular Weight:
330.85 (anhydrous basis)

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing