Skip to Content
MilliporeSigma
Cholesteryl stearate 96%

Cholesteryl stearate

Cholesteryl stearate 96%
Synonym(s):
5-Cholesten-3β-yl octadecanoate
Empirical Formula (Hill Notation):
C45H80O2
CAS Number:
Molecular Weight:
653.12
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing