Skip to Content
MilliporeSigma
Chlorantraniliprole PESTANAL®, analytical standard

Chlorantraniliprole

Chlorantraniliprole PESTANAL®, analytical standard
Synonym(s):
3-Bromo-4′-chloro-1-(3-chloro-2-pyridyl)-2′-methyl-6′-(methylcarbamoyl)pyrazole-5-carboxanilide, 3-Bromo-N-[4-chloro-2-methyl-6-[(methylamino)carbonyl]phenyl]-1-(3-chloro-2-pyridinyl)-1H-pyrazole-5-carboxamide, DPX E2Y45
Empirical Formula (Hill Notation):
C18H14BrCl2N5O2
CAS Number:
Molecular Weight:
483.15

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique