Skip to Content
MilliporeSigma
Cerium(III) oxalate hydrate 99.9% trace metals basis

Cerium(III) oxalate hydrate

Cerium(III) oxalate hydrate 99.9% trace metals basis
Synonym(s):
Cerium oxalate nonahydrate
Linear Formula:
Ce2(C2O4)3 · xH2O
CAS Number:
Molecular Weight:
544.29 (anhydrous basis)
EC Number:

Products

Brand
Formula Weight
Physical Form
Purity
Chemical Composition
Quality Segment
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing