Skip to Content
MilliporeSigma
Cephalomannine ≥97% (HPLC), solid

Cephalomannine

Cephalomannine ≥97% (HPLC), solid
Synonym(s):
NSC 318735
Empirical Formula (Hill Notation):
C45H53NO14
CAS Number:
Molecular Weight:
831.90

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Activity Spectrum
Research Area
Quality Segment
Mode of Action
Showing 1-1 of 1
Compare
Product Number
Description
Pricing