
Cephaëline hydrochloride

Synonym(s):
(−)-Cephaeline dihydrochloride, NSC 32944
Empirical Formula (Hill Notation):
C28H38N2O4 · 2HCl
CAS Number:
Molecular Weight:
539.53
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing