Skip to Content
MilliporeSigma
Carvedilol Related Compound A certified reference material, pharmaceutical secondary standard

Carvedilol Related Compound A

Carvedilol Related Compound A certified reference material, pharmaceutical secondary standard
Synonym(s):
1-{{9-{2-Hydroxy-3-{[2-(2-methoxyphenoxy)ethyl]amino}propyl}-9H-carbazol-4-yl}oxy}-3-{[2-(2-methoxyphenoxy)ethyl]amino}-2-propanol
Empirical Formula (Hill Notation):
C36H43N3O7
CAS Number:
Molecular Weight:
629.74

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade