
Carbon-13C dioxide

Synonym(s):
13C Labeled carbon dioxide, 13CO2
Empirical Formula (Hill Notation):
13CO2
CAS Number:
Molecular Weight:
45.00
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Isotope
Showing 1-6 of 6
Compare
Product Number
Description
Pricing