Skip to Content
MilliporeSigma
Caffeine phyproof® Reference Substance

Caffeine

Caffeine phyproof® Reference Substance
Synonym(s):
1,3,7-Trimethylxanthine, Caffeine
Empirical Formula (Hill Notation):
C8H10N4O2
CAS Number:
Molecular Weight:
194.19
EC Number:

Products

Product Category
Brand
Feature
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Product Line
Purity
Agency
Format
Research Area
Quality Segment
Application
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique