Skip to Content
MilliporeSigma
Butyl acetate SAJ first grade, ≥98.0%

Butyl acetate

Butyl acetate SAJ first grade, ≥98.0%
Linear Formula:
CH3COO(CH2)3CH3
CAS Number:
Molecular Weight:
116.16
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique