Skip to Content
MilliporeSigma
Butorphanol (+)-tartrate salt ≥99%

Butorphanol (+)-tartrate salt

Butorphanol (+)-tartrate salt ≥99%
Synonym(s):
L-N-Cyclobutylmethyl-3,14-dihydroxymorphinan (+)-tartrate salt
Empirical Formula (Hill Notation):
C21H29NO2 · C4H6O6
CAS Number:
Molecular Weight:
477.55
EC Number:

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Research Area
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing