Skip to Content
MilliporeSigma
Bupivacaine hydrochloride British Pharmacopoeia (BP) Reference Standard

Bupivacaine hydrochloride

Bupivacaine hydrochloride British Pharmacopoeia (BP) Reference Standard
Synonym(s):
Bupivacaine hydrochloride monohydrate, 1-Butyl-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide hydrochloride hydrate, 1-Butyl-N-(2,6-dimethylphenyl)-2-piperidinecarboxamide
Empirical Formula (Hill Notation):
C18H28N2O · HCl · H2O
CAS Number:
Molecular Weight:
342.90

Products

Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Technique