Skip to Content
MilliporeSigma
BQ-788 ≥95%, solid

BQ-788

BQ-788 ≥95%, solid
Synonym(s):
2,6-Dimethylpiperidinecarbonyl-γ-Methyl-Leu-Nin-(Methoxycarbonyl)-D-Trp-D-Nle, N-[N-[N-[(2,6-Dimethyl-1-piperidinyl)carbonyl]-4-methyl-L-leucyl]-1-(methoxycarbonyl)-D-tryptophyl]-D-norleucine sodium salt
Empirical Formula (Hill Notation):
C34H50N5NaO7
CAS Number:
Molecular Weight:
663.78

Products

Product Category
Brand
Color
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing