Skip to Content
MilliporeSigma
BQ-123 ≥99%, sodium salt, lyophilized powder

BQ-123

BQ-123 ≥99%, sodium salt, lyophilized powder
Empirical Formula (Hill Notation):
C31H41N6O7 Na
CAS Number:
Molecular Weight:
632.68

Products

Product Category
Brand
Formula Weight
Gene Alias
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing