Skip to Content
MilliporeSigma
Boc-Pro-OH ≥99.0% (T)

Boc-Pro-OH

Boc-Pro-OH ≥99.0% (T)
Synonym(s):
N-(tert-Butoxycarbonyl)-L-proline, Boc-L-proline, Boc-Pro-OH
Empirical Formula (Hill Notation):
C10H17NO4
CAS Number:
Molecular Weight:
215.25

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing