Skip to Content
MilliporeSigma
Boc-Phe(4-I)-OH ≥99.0% (TLC)

Boc-Phe(4-I)-OH

Boc-Phe(4-I)-OH ≥99.0% (TLC)
Synonym(s):
Boc-4-iodo-L-phenylalanine
Linear Formula:
(CH3)3COCONHCH(COOH)CH2C6H4I
CAS Number:
Molecular Weight:
391.20

Products

Brand
Formula Weight
Melting Point (°C)
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing