Skip to Content
MilliporeSigma
Boc-Lys(Boc)-OSu ≥97.0% (calc. based on dry substance, C/N)

Boc-Lys(Boc)-OSu

Boc-Lys(Boc)-OSu ≥97.0% (calc. based on dry substance, C/N)
Synonym(s):
Nα,Nε-Di-Boc-L-lysine hydroxysuccinimide ester
Empirical Formula (Hill Notation):
C20H33N3O8
CAS Number:
Molecular Weight:
443.49

Products

Brand
Formula Weight
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing