Skip to Content
MilliporeSigma
Boc-D-Met-OH ≥98.0% (TLC)

Boc-D-Met-OH

Boc-D-Met-OH ≥98.0% (TLC)
Synonym(s):
Boc-D-methionine, Boc-D-Met-OH
Empirical Formula (Hill Notation):
C10H19NO4S
CAS Number:
Molecular Weight:
249.33

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing