Skip to Content
MilliporeSigma
Boc-Arg(NO2)-OH ≥98.5% (calc. based on dry substance, T)

Boc-Arg(NO2)-OH

Boc-Arg(NO2)-OH ≥98.5% (calc. based on dry substance, T)
Synonym(s):
Nα-Boc-Nω-nitro-L-arginine
Linear Formula:
HN=C(NHNO2)NH(CH2)3CH(COOH)NHCOOC(CH3)3
CAS Number:
Molecular Weight:
319.31

Products

Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Markush Class
Markush Group
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing