Skip to Content
MilliporeSigma
Bisacodyl Related Compound B certified reference material, pharmaceutical secondary standard

Bisacodyl Related Compound B

Bisacodyl Related Compound B certified reference material, pharmaceutical secondary standard
Synonym(s):
2-[( 4-Hydroxyphenyl)-2-pyridinylmethyl]phenol, 2,4′-(Pyridin-2-ylmethylene)diphenol
Empirical Formula (Hill Notation):
C18H15NO2
CAS Number:
Molecular Weight:
277.32

Products

Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade