Skip to Content
MilliporeSigma
BIOshell A400 Protein C4, 3.4 μm Guard Cartridge 3.4 μm particle size, L × I.D. 0.5 cm × 4.6 mm, pkg of 3 ea

BIOshell A400 Protein C4 (3.4 μm) HPLC Columns

BIOshell A400 Protein C4, 3.4 μm Guard Cartridge 3.4 μm particle size, L × I.D. 0.5 cm × 4.6 mm, pkg of 3 ea

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-13 of 13
Compare
Product Number
Description
Pricing