Skip to Content
MilliporeSigma
β-Hydroxypyruvic acid ≥95.0% (dry substance, T)

β-Hydroxypyruvic acid

β-Hydroxypyruvic acid ≥95.0% (dry substance, T)
Synonym(s):
3-Hydroxy-2-oxopropanoic acid
Empirical Formula (Hill Notation):
C3H4O4
CAS Number:
Molecular Weight:
104.06

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing