Skip to Content
MilliporeSigma
β-Hydroxyisovaleric acid ≥95.0% (T)

β-Hydroxyisovaleric acid

β-Hydroxyisovaleric acid ≥95.0% (T)
Synonym(s):
3-Hydroxy-3-methylbutyric acid
Empirical Formula (Hill Notation):
C5H10O3
CAS Number:
Molecular Weight:
118.13

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing