Skip to Content
MilliporeSigma
Bethanechol Chloride Pharmaceutical Secondary Standard; Certified Reference Material

Bethanechol chloride

Bethanechol Chloride Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Carbamyl-β-methylcholine chloride, Bethanechol chloride
Linear Formula:
C7H17N2O2Cl
CAS Number:
Molecular Weight:
196.68

Products

Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Special Grade