Skip to Content
MilliporeSigma
Betadex Sulfobutyl Ether Sodium

Betadex sulfobutyl ether sodium

Betadex Sulfobutyl Ether Sodium
Synonym(s):
β-Cyclodextrin sulfobutyl ethers sodium salts
CAS Number:

Products

Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2