Skip to Content
MilliporeSigma
Benzoylecgonine-D8 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®

Benzoylecgonine-D8 solution

Benzoylecgonine-D8 solution 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Empirical Formula (Hill Notation):
C16H11NO4D8
CAS Number:
Molecular Weight:
297.38

Products

Brand
Formula Weight
Manufacturer
Physical Form
Format
Application
Package Size
Reference Material Type
Special Grade
Technique