Skip to Content
MilliporeSigma
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate 97%

(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate

(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate 97%
Synonym(s):
BOP, BOP Reagent, Castros reagent
Empirical Formula (Hill Notation):
C12H22F6N6OP2
CAS Number:
Molecular Weight:
442.28
EC Number:

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Application
Reaction Type
Showing 1-1 of 1