Skip to Content
MilliporeSigma
Benzaldehyde SAJ special grade, ≥98.0%

Benzaldehyde

Benzaldehyde SAJ special grade, ≥98.0%
Synonym(s):
Bitter almond, Benzaldehyde
Linear Formula:
C6H5CHO
CAS Number:
Molecular Weight:
106.12
EC Number:

Products

Product Category
Brand
Sustainability
Boiling Point (°C)
Feature
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique