Skip to Content
MilliporeSigma
Baythroid® PESTANAL®, analytical standard

Baythroid®

Baythroid® PESTANAL®, analytical standard
Synonym(s):
Cyfluthrin
Empirical Formula (Hill Notation):
C22H18Cl2FNO3
CAS Number:
Molecular Weight:
434.29
EC Number:

Products

Product Category
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing