Skip to Content
MilliporeSigma
Azamethiphos PESTANAL®, analytical standard

Azamethiphos

Azamethiphos PESTANAL®, analytical standard
Synonym(s):
S-((6-Chloro-2-oxooxazolo[4,5-b]pyridin-3(2H)-yl)methyl) O,O-dimethyl phosphorothioate
Empirical Formula (Hill Notation):
C9H10ClN2O5PS
CAS Number:
Molecular Weight:
324.68
EC Number:

Products

Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique