Skip to Content
MilliporeSigma
Aspartame European Pharmacopoeia (EP) Reference Standard

Aspartame

Aspartame European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Asp-Phe methyl ester, N-(L-α-Aspartyl)-L-phenylalanine methyl ester, Asp-Phe-OMe, Aspartame, L-Aspartyl-L-phenylalanine methyl ester, N-L-α-Aspartyl-L-phenylalanine methyl ester
Linear Formula:
HOOCCH2CH(NH2)CONHCH(CH2C6H5)COOCH3
CAS Number:
Molecular Weight:
294.30

Products

Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Purity
Agency
Format
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
API Family
Reference Material Type
Special Grade
Technique