Skip to Content
MilliporeSigma
Asialoganglioside GM1 from bovine brain ~98%, lyophilized powder

Asialoganglioside GM1 from bovine brain

Asialoganglioside GM1 from bovine brain ~98%, lyophilized powder
Synonym(s):
Ceramide tetrahexoside, Ganglioside GM1, asialo, Gangliotetraosyl ceramide
CAS Number:

Products

Product Category
Brand
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing