Skip to Content
MilliporeSigma
Ascentis® Phenyl Supelguard Guard Cartridge Kit, 5 μm particle size, L × I.D. 2 cm × 4 mm

Ascentis® Phenyl (5 µm) HPLC Columns

Ascentis® Phenyl Supelguard Guard Cartridge Kit, 5 μm particle size, L × I.D. 2 cm × 4 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Quality Segment
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique