Skip to Content
MilliporeSigma
Ascentis® Express ES-Cyano, 5 μm HPLC Column 5 μm particle size, L × I.D. 5 cm × 3 mm

Ascentis® Express 90 Å ES-Cyano (5 μm) HPLC Columns

Ascentis® Express ES-Cyano, 5 μm HPLC Column 5 μm particle size, L × I.D. 5 cm × 3 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Length (mm)
Quality Segment
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-18 of 18
Compare
Product Number
Description
Pricing