Skip to Content
MilliporeSigma
Ascentis® Express C8 Capillary, 2.7 μm HPLC Column 2.7 μm particle size, L × I.D. 5 cm × 300 μm

Ascentis® Express 90Å C8 (2.7 μm) HPLC Columns

Ascentis® Express C8 Capillary, 2.7 μm HPLC Column 2.7 μm particle size, L × I.D. 5 cm × 300 μm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Length (mm)
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-20 of 21
Page 1 of 2
Compare
Product Number
Description
Pricing
Page 1 of 2