Skip to Content
MilliporeSigma
Ascentis® C18 HPLC Column 5 μm particle size, L × I.D. 10 cm × 2.1 mm

Ascentis® C18 (5 µm) HPLC Columns

Ascentis® C18 HPLC Column 5 μm particle size, L × I.D. 10 cm × 2.1 mm

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-13 of 13
Compare
Product Number
Description
Pricing