Skip to Content
MilliporeSigma
Ascentis® C18 Supelguard Guard Cartridge 3 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 2 ea

Ascentis® C18 (3 µm) HPLC Columns

Ascentis® C18 Supelguard Guard Cartridge 3 μm particle size, L × I.D. 2 cm × 4 mm, pkg of 2 ea

Products

Product Category
Brand
pH Value
Product Line
Agency
USP Suitability
Separation Technique
ID (mm)
Length (cm)
Application
Column Material
Package Size
Particle Size (µm)
Pore Size (Å)
Technique
Showing 1-11 of 11