Skip to Content
MilliporeSigma
Anisole Pharmaceutical Secondary Standard; Certified Reference Material

Anisole

Anisole Pharmaceutical Secondary Standard; Certified Reference Material
Synonym(s):
Methoxybenzene, Methyl phenyl ether, Anisole
Linear Formula:
CH3OC6H5
CAS Number:
Molecular Weight:
108.14
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Format
Quality Segment
Application
Organoleptic Class
Reference Material Type
Special Grade
Technique