Skip to Content
MilliporeSigma
Amygdalin phyproof® Reference Substance

Amygdalin

Amygdalin phyproof® Reference Substance
Synonym(s):
D-Mandelonitrile 6-O-β-D-glucosido-β-D-glucoside, D-Mandelonitrile-β-gentiobioside, Amygdaloside, Laetrile
Empirical Formula (Hill Notation):
C20H27NO11
CAS Number:
Molecular Weight:
457.43

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Purity
Format
Tissue
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Technique
Showing 1-3 of 3