Skip to Content
MilliporeSigma
Ammonium sodium phosphate dibasic tetrahydrate BioXtra, ≥99%

Ammonium sodium phosphate dibasic tetrahydrate

Ammonium sodium phosphate dibasic tetrahydrate BioXtra, ≥99%
Synonym(s):
sec-Sodium ammonium phosphate, Ammonium sodium hydrogen phosphate, Sodium ammonium hydrogen phosphate
Linear Formula:
NaNH4HPO4 · 4H2O
CAS Number:
Molecular Weight:
209.07

Products

Product Category
Formula Weight
Physical Form
Product Line
Purity
Special Grade
Showing 1-2 of 2