Skip to Content
MilliporeSigma
Ammonium hydroxide solution BioUltra, ~1 M NH3 in H2O (T)

Ammonium hydroxide solution

Ammonium hydroxide solution BioUltra, ~1 M NH3 in H2O (T)
Synonym(s):
Ammonia aqueous, Ammonia water
Linear Formula:
NH4OH
CAS Number:
Molecular Weight:
35.05

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Agency
Quality Segment
Special Grade