
Amiodarone Related Compound E

Synonym(s):
(2-Butylbenzofuran-3-yl)(4-hydroxyphenyl)methanone, NSC 85438
Empirical Formula (Hill Notation):
C19H18O3
CAS Number:
Molecular Weight:
294.34
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2