Skip to Content
MilliporeSigma
Allyltributylstannane 97%

Allyltributylstannane

Allyltributylstannane 97%
Synonym(s):
Allyltri-n-butyltin, Allyltributyltin, Tributyl-2-propenylstannane
Linear Formula:
CH2=CHCH2Sn(CH2CH2CH2CH3)3
CAS Number:
Molecular Weight:
331.12
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing