Skip to Content
MilliporeSigma
Allyl sulfide 97%

Allyl sulfide

Allyl sulfide 97%
Synonym(s):
3,3′-Thiobis-1-propene, Diallyl sulfide, NSC 20947
Linear Formula:
(CH2=CHCH2)2S
CAS Number:
Molecular Weight:
114.21
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing