Skip to Content
MilliporeSigma
vibrant-m

Allopurinol impurity B

vibrant-m
Synonym(s):
3-(Formylamino)-1H-pyrazole-4-carboxamide, 5-(Formylamino)-1H-pyrazole-4-carboxamide
Empirical Formula (Hill Notation):
C5H6N4O2
CAS Number:
Molecular Weight:
154.13

Products

Formula Weight
Manufacturer
Physical Form
Format
Application
API Family
Reference Material Type
Special Grade