Skip to Content
MilliporeSigma
Acriflavine suitable for fluorescence, BioReagent, ≥90% (AT)

Acriflavine

Acriflavine suitable for fluorescence, BioReagent, ≥90% (AT)
Synonym(s):
3,6-Diamino-10-methylacridinium chloride mixt. with 3,6-diaminoacridine (proflavine), Euflavine, Trypaflavine Neutral
Empirical Formula (Hill Notation):
C14H14ClN3
CAS Number:
Molecular Weight:
259.73

Products

Product Category
Brand
Formula Weight
Physical Form
Product Line
Purity
Chemical Composition
Quality Segment
Application
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing