Skip to Content
MilliporeSigma
Acridine Orange hemi(zinc chloride) salt Dye content 85 %

Acridine Orange hemi(zinc chloride) salt

Acridine Orange hemi(zinc chloride) salt Dye content 85 %
Synonym(s):
3,6-Bis(dimethylamino)acridine hydrochloride zinc chloride double salt, 3,6-Bis(dimethylamino)acridinium chloride hemi(zinc chloride salt), Basic Orange 14
Linear Formula:
C17H20ClN3 · HCl · 1/2ZnCl2
CAS Number:
Molecular Weight:
369.96
EC Number:

Products

Product Category
Formula Weight
Physical Form
Product Line
Chemical Composition
Quality Segment
Application
Technique